Learn More
Invitrogen™ IP-10 (CXCL10) Human ELISA Development Kit (TMB), PeproTech®, PeproTech®
ELISA
Brand:  Invitrogen™ 900-T39
Additional Details : Weight : 0.03500kg
Product Code. 17819195
  • 330.00€ / Pack of 10
Estimated Shipment: 11-06-2024
Description
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
Specifications
Specifications
P02778
3627
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
ELISA
CXCL10