Learn More
HYDRA-POINT™ Coulometric Generator Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Brand: JT Baker 6281-00
114.00 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Additional Details : Weight : 0.16000kg Transport : UN number : 1188 Chem class : 3 Pack group : III
Description
- For Karl Fischer titration
Specifications
46.00°C | |
HYDRA-POINT™ Coulometric Generator Solution | |
BAKER ANALYZED™ Reagent |
125 mL | |
Narrow Mouth Amber Glass Bottle |
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.