Learn More
Cytiva HyClone™ SFM4HEK293™ Media
36.40€ - 80.45€
Specifications
Certifications/Compliance | cGMP |
---|---|
Description | SFM4HEK293 Media |
Form | Liquid |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10500283
|
Cytiva
SH30521.02 |
SFM4HEK293 Media | 1000 mL bottle |
80.45€
1000mL |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
10061502
|
Cytiva
SH30521.01 |
SFM4HEK293 Media | 500 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
Support production of adenoviral vectors and recombinant proteins in HEK 239 cultures with HyClone™ SFM4HEK293™ Media, a versatile, protein-free cell culture media containing no animal-derived components.
Specifications
cGMP | |
SFM4HEK293 Media | |
Liquid |