missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ACE2 (aa18-740), His-tag Recombinant Protein

HEK293 cells Recombinant Protein

Brand:  Invitrogen™ RP87672

Additional Details : Weight : 0.01000kg

Product Code. 17174589

  • 596.00€ / 100µg
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.
Specifications

Specifications

Q9BYF1
PBS with no preservative; pH 7.2
Human ACE2 (aa18-740) Fc Chimera
HEK293 cells
RUO
ACE2
Unconjugated
Liquid
Control, ELISA, Western Blot
59272
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885
Human
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers