missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ HEPES, 1.0M buffer soln., pH 8.5
85.30€ - 112.00€
Chemical Identifiers
| CAS | 7365-45-9 |
|---|---|
| Molecular Formula | C8H18N2O4S |
| Molecular Weight (g/mol) | 238.302 |
| MDL Number | MFCD00006158 |
| InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
| PubChem CID | 23831 |
| ChEBI | CHEBI:42334 |
| IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid |
| SMILES | C1CN(CCN1CCO)CCS(=O)(=O)O |
Chemical Identifiers
| 7365-45-9 | |
| 238.302 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| CHEBI:42334 | |
| C1CN(CCN1CCO)CCS(=O)(=O)O |
| C8H18N2O4S | |
| MFCD00006158 | |
| 23831 | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid |
Specifications
| HEPES buffer soln. | |
| C8H18N2O4S | |
| 1.0M buffer soln. | |
| C1CN(CCN1CCO)CCS(=O)(=O)O | |
| 238.302 | |
| CHEBI:42334 | |
| 8.5 |
| 7365-45-9 | |
| MFCD00006158 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid | |
| 23831 | |
| Liquid | |
| 250mL |