Learn More
Fisherbrand™ GT1 Benchtop Centrifuge Packs
Fisherbrand™ Benchtop Centrifuges combine a compact size with operational flexibility, making them the ideal choice for molecular biology laboratories. Available in ventilated or refrigerated versions.
4140.00€ - 6945.00€
Specifications
Certifications/Compliance | IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1, UL Listed, CE marked, Certified biocontainment |
---|---|
Capacity | 4 x 145 mL |
Max. RCF | 3,046 x g |
Max. Speed | 4,350 rpm |
Drive System | Direct Brushless Induction |
Product Code | Brand | Model | Refrigerated | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Refrigerated | Price | Quantity & Availability | ||||
15868722
|
Fisherbrand™
|
GT1R | Yes |
6945.00€
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||||
15878722
|
Fisherbrand™
|
GT1 | No |
4140.00€
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
||||
Description
- Compact size with minimal benchtop footprint
- Ventilated and refrigerated versions available, with rapid temperature pull-downs
- Includes TX-150 swing-out rotor with adapters accommodating 4x50mL or 8x15mL conical tubes
- One-touch, easy-to-use operation with pre-saved protocols
- Backlit display interface with multi-language options
- Rapid push-button rotor exchange with the Auto-Lock™ rotor swap system
- Corrosion resistant and lubricating rotor-cross coating
- Fitted with ClickSeal™ biocontainment lids
- Optional high-speed microlitre rotor also available for molecular biology applications
- Glove and wipe-clean, detergent-friendly
- Two year warranty
Specifications
IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1, UL Listed, CE marked, Certified biocontainment | |
3,046 x g | |
Direct Brushless Induction | |
Four direct access programs | |
Yes | |
ClickSeal biocontainment lid | |
2 Years (Centrifuge), 5 Years (Rotor) | |
50/60 Hz |
4 x 145 mL | |
4,350 rpm | |
TX-150 rotor, ClickSeal lid, four adapters for 4x50mL conical tubes, four adapters for 8x15mL conical tubes | |
TX-150 | |
10 sec. to 99 hr. 59 min. | |
Benchtop Centrifuge | |
230 V |