Learn More
IgG (H+L) Goat anti-Rabbit, AP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31340
Additional Details : Weight : 0.00100kg Transport : UN number : 3265 Chem class : 8 Pack group : II
Description
Thermo Scientific Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Rabbit IgG (H+L) | |
| Polyclonal | |
| AP | |
| Lyophilized | |
| Purified Rabbit IgG, whole molecule | |
| Affinity chromatography | |
| RUO | |
| Secondary Antibody | |
| Polyclonal |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 0.6 mg/mL | |
| TBS with 15mg/mL BSA and no preservative; pH 8 | |
| Goat | |
| IgG | |
| 1 mL | |
| 4° C | |
| Secondary | |
| Rabbit |