Learn More
Socorex™ Glass Barrels
90.60€ - 111.00€
Specifications
Product Type | Glass Barrel |
---|---|
For Use With (Equipment) | Self-refilling Syringe |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10560753
|
Socorex™
0187.10 |
10 mL |
111.00€
Pack of 6 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
10151872
|
Socorex™
0187.02 |
2 mL |
90.60€
Pack of 6 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
Specifications
Glass Barrel | |
Self-refilling Syringe |