missing translation for 'onlineSavingsMsg'
Learn More

Fusidic acid, 98%, Thermo Scientific Chemicals

Product Code. p-7077732 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15542333 1 g 1g
15552333 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15542333 Supplier Thermo Scientific Acros Supplier No. 458460010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6990-06-3
ChEBI CHEBI:29013

Specifications

Infrared Spectrum Authentic
Molecular Formula C31H48O6
Quantity 1 g
Synonym fusidic acid, fusidine, ramycin, fucithalmic, fucidic acid, fucidin acid, fusidate, fucidin, flucidin, fucidate
InChI Key IECPWNUMDGFDKC-MZJAQBGESA-N
SMILES CC1C2CCC3(C(C2(CCC1O)C)C(CC4C3(CC(C4=C(CCC=C(C)C)C(=O)O)OC(=O)C)C)O)C
IUPAC Name (2Z)-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16S)-16-acetyloxy-3,11-dihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid
Molecular Weight (g/mol) 516.72
PubChem CID 3000226
Formula Weight 516.71
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Fusidic acid
Show More Show Less
compliance-icons
Product Identifier
  • Fusidic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.