Learn More
Eppendorf™ Fixed Angle Rotors for Concentrator Plus Centrfuge
For use with Concentrator Plus Centrifuge
457.00€
Specifications
Type | Fixed Angle Rotor |
---|---|
For Use With (Equipment) | Concentrator Plus Centrifuge |
Angle | Fixed |
Includes | Without Lid |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
10504843
|
Eppendorf™
5490030001 |
48 x 1.5/2 mL |
457.00€
Each |
In Stock
Log in to see stock. |
|||||
10184313
|
Eppendorf™
5490034007 |
72 x 0.5 mL |
457.00€
Each |
In Stock
Log in to see stock. |
|||||
10572264
|
Eppendorf™
5490035003 |
36 x 3.0/5.0 mL Flat-bottom Tubes |
457.00€
Each |
In Stock
Log in to see stock. |
|||||
10115802
|
Eppendorf™
5490032004 |
70 x 1.5/2 mL |
457.00€
Each |
In Stock
Log in to see stock. |
|||||
Description
- Eppendorf Concentrator plus was designed with the customer in mind to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench.
Specifications
Fixed Angle Rotor | |
Concentrator Plus Centrifuge | |
Fixed | |
Without Lid |