missing translation for 'onlineSavingsMsg'
Learn More

UVP Filter excitation for automated BioLite 600nm to 645nm UVP

Brand:  UVP 38-0359-04

Additional Details : Weight : 0.20000kg

Product Code. 12853079

  • 670.00€ / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket