Learn More
UVP Filter excitation for automated BioLite 280nm to 450nm UVP
Brand:  UVP 38-0358-04
Additional Details : Weight : 0.20000kg
Product Code. 12813079
  • 670.00€ / Each
Estimated Shipment: 02-08-2024