Learn More
Thermo Scientific™ T7 RNA Polymerase (20 U/μL)
Product Code. p-4530777 Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
20U/µL
Quantity:
5 x 5,000 units
5,000 units
Unit Size:
25000 units
5000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration Quantity unitSize
10610301 20U/µL 5,000 units 5000 units
10212610 20U/µL 5 x 5,000 units 25000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10610301 Supplier Thermo Scientific™ Supplier No. EP0111
Thermo Scientific™ T7 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.
Thermo Scientific™ Bacteriophage T7 RNA polymerase is DNA-dependent RNA polymerase with strict specificity its respective double-stranded promoter.
It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used:for hybridization, in vitro RNA translation; as aRNA, siRNA, substrate in RNase protection assays, template for genomic DNA sequencing.
Note:
Consensus promoter sequences: TAATACGACTCACTATA G GGAGA. The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively.
TRUSTED_SUSTAINABILITY
Specifications
Polymerase T7 RNA Polymerase
Quantity 5,000 units
Concentration 20U/µL