Learn More
Thermo Scientific™ T4 DNA Polymerase (5 U/μL)
Product Code. p-4521340 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Units
500 Units
Unit Size:
100 units
500 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10680281 100 Units 100 units
10531051 500 Units 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10680281 Supplier Thermo Scientific™ Supplier No. EP0061
Ensure accuracy in DNA synthesis with template-dependent T4 DNA Polymerase that catalyzes the 5'-3' synthesis from primed single-stranded DNA.
T4 DNA Polymerase, a template-dependent DNA polymerase, catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.
Highlights
  • Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I, E. coli,and Klenow fragment
  • Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase buffers
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5 U/μL
Polymerase T4 DNA Polymerase
Quantity 100 Units
Includes 0.35 mL of 5X Reaction Buffer