Learn More
Thermo Scientific™ GeneRuler™ DNA Ladder Mix
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.
106.00€ - 418.00€
Specifications
Quantity 5 × 50μg
View More Specs
Products 2
Product Code Brand Kit Contents Price Quantity & Availability  
Product Code Brand Kit Contents Price Quantity & Availability  
11873963 Thermo Scientific™
SM0331
6X DNA Loading Dye, 2 x 1mL 106.00€
Pack of 5 View alternative products
Save Up to 
»
EU Stock 151
Add to basket
11883963 Thermo Scientific™
SM0332
6X DNA Loading Dye, 10 x 1mL 418.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 30
Add to basket
Description
Description
Highlights
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands
  • Supplied with loading dye for sample DNA
Specifications
Specifications
5 × 50μg
This product or its use is covered by at least one claim of U.S. Patent Nos. 5,834,201, 6,680,378, and/or 7,132,520 owned by Invitrogen Corporation. The purchase of this product conveys to the buyer the non-transferable right to use the purchased amount of the product in internal research conducted by the buyer. The buyer cannot use this product or materials made by the employment of this product for Commercial Purposes. Commercial Purposes means any activity for consideration and may include, but is not limited to: (1) use of the product in manufacturing; (2) use of the product to provide a service, information, or data; (3) use of the product for therapeutic, diagnostic or prophylactic purposes; or (4) resale of the product, whether or not the product is resold for use in research. Further information on purchasing licenses under the above patents may be obtained by contacting the Business Development Department, Invitrogen Corporation, 5791 Van Allen Way, Carlsbad, CA 92008.