missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETFDH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | ETFDH |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16847963
|
Proteintech
11109-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16837963
|
Proteintech
11109-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This antibody is specific to ETFDH.
Electron-transferring-flavoprotein dehydrogenase in the inner mitochondrial membrane accepts electrons from electron-transfer flavoprotein which is located in the mitochondrial matrix and reduces ubiquinone in the mitochondrial membrane. The protein is synthesized as a 67-kDa precursor which is targeted to mitochondria and processed in a single step to a 64-kDa mature form located in the mitochondrial membrane. Deficiency in electron-transferring-flavoprotein dehydrogenase have been demonstrated in some patients with type II glutaricacidemia.Specifications
ETFDH | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q16134, Q6UPE1, Q921G7 | |
2110, 295143, 66841 | |
ETFDH Fusion Protein Ag1568 | |
Primary | |
-20°C | |
ETFDH |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ETF dehydrogenase, ETF QO, ETF ubiquinone oxidoreductase, ETFDH, ETFQO, MADD | |
Etfdh | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |