Learn More
JP Selecta™ Dur-Aluminum Rod
10.44€ - 54.00€
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12192952
|
JP Selecta™
6000798 |
500 mm |
54.00€
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12087874
|
JP Selecta™
6000270 |
700 mm |
10.44€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
Specifications
Dur-Aluminum | |
Rod | |
1 Ea. |