Learn More
Socorex™ Dosys Stainless Steel Dispensing Cannula
5.45€ - 32.00€
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Stainless Steel |
Product Type | Dispensing Cannula |
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10039502
|
Socorex™
370.0840 |
0.8 mm |
32.00€
Pack of 12 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
10282503
|
Socorex™
370.1250 |
1.2 mm |
5.45€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
Specifications
Autoclavable | |
Stainless Steel | |
Dispensing Cannula |