missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DKC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | DKC1 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883995
|
Proteintech
25420-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16873995
|
Proteintech
25420-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
May function as a transcriptional repressor. May also function as a ubiquitin ligase mediating ubiquitination of chromatin-associated TP53. Functions in cell survival and proliferation through control of the cell cycle. Functions in the p53 and pRB tumor suppressor pathways and regulates the cyclin CCNA2 transcription.; FUNCTION: Identified as a cellular target of the adenoviral oncoprotein E1A, it is required for both transcriptional activation and repression of viral genes.Specifications
DKC1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O60832 | |
1736 | |
DKC1 Fusion Protein Ag7992 | |
Primary | |
-20°C | |
DKC1 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CBF5, CBF5 homolog, DKC, DKC1, Dyskerin, NAP57, NOLA4, Nucleolar protein NAP57, snoRNP protein DKC1, XAP101 | |
DKC1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |