missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: Near C-terminus
Immunogen: A synthetic peptide mapping near the C-terminus of human desmin
Molecular Weight: 53 kDa
Positive Control: Muscle or Sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
Specifications
| Antigen | Desmin (Muscle Cell Marker) |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide mapping near the C-terminus of human desmin |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Research Discipline | Muscle Biology |
| Show More |
Safety and Handling
Product Identifier
- 500 UL DESMIN (MUSCLE CELL MARKER) EPITOPE SPECIFI c Rabbit antibody (Purified Ab with BSAand Azide) CE IVD, Isotype: IgG, Species reactivity: Hu, Ms, Rt, Application: IH, IP,WB200µg/mL 0.5mL store at 2° to 8°C
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H330-Fatal if inhaled.
- H370-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P320-Specific treatment is urgent (see on this label).
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- MIXTURE LIST-Contain: methanol, glycerin,N-ethyl-ethanamine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?