missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin.
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: Near C-terminus
Immunogen: A synthetic peptide mapping near the C-terminus of human desmin
Molecular Weight: 53 kDa
Positive Control: Muscle or Sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
Specifications
| Antigen | Desmin (Muscle Cell Marker) |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide mapping near the C-terminus of human desmin |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?