Learn More
Fisherbrand™ Crimping Heads for Pneumatic Hand-held Crimping Tool
For use with Pneumatic Hand-held Crimping Tool.
Brand:  Fisherbrand™
1225.50 EUR valid until 2024-12-31
Use promo code "23262" to get your promotional price.
Additional Details : Weight : 0.76000kg
 View more versions of this product
Product Code. 11879440
  • 1290.00€ / Each
Estimated Shipment: 18-06-2024
Description
  • Crimping Head
  • Pneumatic
  • Steel
Specifications
Specifications
Flip Top/Flip Off Seals
32mm Crimping Head