Learn More
B Braun™ Combi-Stopper Closing Cones
B Braun™ Combi-Stopper Closing Cones are suitable for sterile closings of IV and syringe assemblies
16.25€
Specifications
Material | Polyethylene |
---|---|
Product Type | Combi Stopper |
Connection Type | Luer Lock |
Sterility | Sterile |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10712393
|
B Braun™
4495152 |
Blue |
16.25€
Pack of 100 |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
10500053
|
B Braun™
4495101 |
Red |
16.25€
Pack of 100 |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
Description
- Luer Lock fitting male and female
- Made of PE (Polyethylene)
- Sterile closing of all kinds of open IV-accesses in IV-Sets
- Sterile closing of pre-filled syringes
- Available in red, blue and white
- Box of 100 pcs
Specifications
Polyethylene | |
Combi Stopper | |
Luer Lock | |
Sterile |