missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen Type VI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifications
Antigen | Collagen Type VI |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836834
|
Proteintech
17023-1-AP-20UL |
20 μL |
140.00€
20µL |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16826834
|
Proteintech
17023-1-AP-150UL |
150 μL |
399.00€
150µL |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
The extensive family of COL gene products(collagens) iscomposed ofseveral chain types, including fibril-forming interstitialcollagens(typesI, II, III and V) and basement membrane collagens(type IV), each type containing multiple isoforms. Collagens are fibrous, extracellularmatrix proteinswith high tensile strength and are themajorcomponents ofconnective tissue,such astendons and cartilage. Allcollagenscontain a triple helix domain and frequentlyshow lateralself-association in order to formcomplexconnective tissues. Several collagens also play a role in cell adhesion, important formaintaining normal tissue architecture and function.Specifications
Collagen Type VI | |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P12109, Q04857 | |
12833, 1291, 294337 | |
Collagen Type VI Fusion Protein Ag10288 | |
Primary | |
-20°C | |
COL6A1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COL6A1, collagen 6A1, Collagen alpha 1(VI) chain, collagen, type VI, alpha 1, OPLL | |
COL6A1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |