Learn More
Proteintech ChromoTek pSpot-Tag-Actin Vector
pSpot Spot-Tag™-Actin vector for expression of Spot-Tag™ β-actin fusion protein in mammalian cells - positive control.
Brand: Proteintech EV-31
Additional Details : Weight : 0.00240kg
Description
- The plasmid pSpot-Tag™-Actin is a mammalian expression vector encoding human β-actin fused to the Spot-Tag™. The Spot-Tag™ comprises the sequence PDRVRAVSHWSS and is fused to the N-terminus of β-actin. This plasmid may be used as a positive control for immunofluorescence microscopy or Western blot experiments using the Spot-Tag™ system.
Specifications
10 μg |