Learn More
Macherey-Nagel™ CHROMABOND™ Flash RS SiOH Cartridges
Ideal for Flash separations from 10mg up to 300g. Macherey-Nagel™ CHROMABOND™ Flash RS SiOH Cartridges are pre-packed columns for direct use in Teledyne Isco™ systems. Available in a range of sizes with 25–300g of SiOH adsorbent.
188.00€ - 449.00€
Specifications
Material | Polypropylene |
---|---|
Style | Female Luer lock inlet, male Luer outlet |
Type | Flash RS SiOH Column |
Column Type | Flash Column |
For Use With (Application) | Flash Chromatography |
Product Code | Brand | Includes | Length (Metric) | Diameter (Metric) Outer | Max. Pressure | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Length (Metric) | Diameter (Metric) Outer | Max. Pressure | Price | Quantity & Availability | ||
11653109
|
Macherey-Nagel™
732802 |
25 g SiOH | 165 mm | 21.2 mm ID | 21 bar |
188.00€
Pack of 15 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
12768322
|
Macherey-Nagel™
732806 |
200 g SiOH | 200 mm | 60.0 mm ID | 12 bar |
449.00€
Pack of 6 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
12853961
|
Macherey-Nagel™
732805 |
120 g SiOH | 255 mm | 36.0 mm ID | 15 bar |
311.00€
Pack of 10 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
12758322
|
Macherey-Nagel™
732804 |
80 g SiOH | 240 mm | 30.8 mm ID | 15 bar |
312.00€
Pack of 12 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||
Description
For convenient operation and reliable upscaling, these ready-to-use Flash cartridges can be used with the Isco Companion™ and other Teledyne Isco CombiFlash™ systems, or as a stand-alone version for all pump/detector combinations (e.g., from Biotage™, Büchi™)
- Economical, ready to use cartridges increase flexibility and save time and money
- Increases analytical safety—high pressure stability, excellent separation efficiency, good reproducibility
- Packed with 25g to 200g SiOH adsorbent
- Corresponding TLC plates: silica layers
- Macherey-Nagel™ is one of the leading companies in silica and TLC
Specifications
Polypropylene | |
Flash RS SiOH Column | |
Flash Chromatography |
Female Luer lock inlet, male Luer outlet | |
Flash Column | |
Directly Used in The Teledyne Isco Companion, Rf |