Learn More
Invitrogen™ Celleste™ 2D Deconvolution Module, network license (includes one seat)
The Celleste 5.0 Image Analysis package easily transforms images into quantitative data in a streamlined workflow. The Celleste Celleste 2D Deconvolution Module for Network License helps improve single-plane image quality of cells or tissue slices by clearing background haze
Brand:  Invitrogen™ AMEP4915
Additional Details : Weight : 0.10000kg
Product Code. 16678977
  • 2565.00€ / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
The Celleste 5.0 Image Analysis package easily transforms images into quantitative data in a streamlined workflow. The Celleste 2D Deconvolution Module for Network License helps improve single-plane image quality (S/N) of cells or tissue slices by clearing background haze (out-of-focus light). Coupled with the powerful image acquisition capabilities of the EVOS microscopes, Celleste 5 software allows you to seamlessly capture, process, measure, analyze and share images and data. A Network Server License is required for use with this module.
Celleste 5.0 Image Analysis Software Network License [1] is a full-feature image analysis package designed for a range of biological applications, from image adjustments and processing with manual and automatic measurements over multiple channels, to segmentation and classification tools that help you transform images into quantitative data in a streamlined and customizable workflow. The Celleste 5.0 Image Analysis Network Server License is designed for environments where the use of multiple copies is desired. The Network Server License comes with one seat and additional Soft License Seats [2] can be added at any time. Optional modules for 2D and 3D deconvolution, visualization, analysis and GPU-powered processing allow you to customize the capabilities according to your needs. The Celleste 2D Deconvolution Module for Network License helps improve single-plane image quality (S/N) of cells or tissue slices by clearing background haze (out-of-focus light).Celleste 5.0 software with Celleste 2D Deconvolution Module for Network License offers you these important advantages:• Streamlined workflow tools that make it easy to visualize and edit images• Powerful image analysis capabilities for segmentation, classification, and quantification of single images or a batch of images• 2D blind and non-blind deconvolution options• Dramatically improved image quality• Removal of blur that can obscure important details• The tools to generate more impressive images and reliable dataMore about the deconvolution modulesDeconvolution is an algorithm-based process used to restore systematic loss of image contrast (blur) that results from the image point spread function (PSF). The powerful 2D and 3D restoration algorithms available in the Celleste 5 modules allow you to deconvolve your images to enhance contrast and quality, or to visualize them in Z, over time and by channel, then analyze the resulting image using an expanded tool set.[1] Celleste 5.0 Image Analysis Network Server License [AMEP4868][2] Celleste Image Analysis Network Seat [AMEP4869]Explore the entire EVOS line of imaging systems, accessories, and offers [3] [www.thermofisher.com/EVOS]For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Celleste
EVOS M5000, EVOSM7000 and Other Imaging Systems