missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin G Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA579097

Additional Details : Weight : 0.01000kg

Product Code. 15994665

  • 421.00€ / 100µg
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.
Specifications

Specifications

Cathepsin G
Polyclonal
Unconjugated
CTSG
CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP
Rabbit
Antigen affinity chromatography
RUO
1511, 290257
-20°C
Lyophilized
Western Blot
500 μg/mL
PBS with 5MG BSA and 0.05MG sodium azide
P08311, P17977
CTSG
E.coli-derived human Cathepsin G recombinant protein (Position: A76-L255).
100 μg
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers