Learn More
Thermo Scientific Chemicals cAMP Dependent PK Inhibitor, PKI (14-24), Amide
For cellular and molecular biology applications
Brand:  Thermo Scientific Chemicals J66795.M¬
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15418299
  • 107.00€ / 1mg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
HPLC
Solid
cAMP Dependent PK Inhibitor, PKI (14-24), amide
RUO - research use only
-30°C to -10°C
1251.37
1 mg
Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp-NH2
Product Suggestions
Product Suggestions
Videos
SDS Documents