Learn More
Pal International™ Bouffant Caps
77.72 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Color:
White
Description
- Soft, light non-woven polypropylene with an elasticated edge
- Complete hair coverage
- Ultra low-linting, making them ideally suited to working environments where contamination by loose fibers may be a risk
- Food safe and Latex-free for reduced irritation
- Larger than a mob cap and designed for longer hair
- One size fits all
Specifications
Product Type | Bouffant |
Size | One Size |
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Latex Free | Yes |
Model | D84110 |
Quantity | 1000/Pk. |