Learn More
BRAND™ Polypropylene Test Tube Racks
Product Code. p-3361221 Shop All Brand Products
Change view
Click to view available options
Color:
Blue
Red
White
Yellow
Holds:
Tubes (13 mm)
Tubes (16 mm)
Tubes (18 mm)
Tubes (20 mm)
Tubes (25 mm)
Tubes (30 mm)
Unit Size:
Pack of 5
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Holds unitSize
10603721 White Tubes (13 mm) Pack of 5
10014070 Blue Tubes (13 mm) Pack of 5
11804180 Red Tubes (13 mm) Pack of 5
10666511 Yellow Tubes (13 mm) Pack of 5
11814040 White Tubes (18 mm) Pack of 5
10465421 Blue Tubes (18 mm) Pack of 5
10363701 Red Tubes (18 mm) Pack of 5
10654291 Yellow Tubes (18 mm) Pack of 5
10088190 White Tubes (20 mm) Pack of 5
10271951 Blue Tubes (20 mm) Pack of 5
10150591 Red Tubes (20 mm) Pack of 5
10028180 Yellow Tubes (20 mm) Pack of 5
10175630 White Tubes (25 mm) Pack of 5
10364081 Blue Tubes (25 mm) Pack of 5
10359760 Red Tubes (25 mm) Pack of 5
11894170 Yellow Tubes (25 mm) Pack of 5
11894150 White Tubes (30 mm) Pack of 5
11804160 Blue Tubes (30 mm) Pack of 5
11814160 Red Tubes (30 mm) Pack of 5
11824160 Yellow Tubes (30 mm) Pack of 5
11889550 White Tubes (16 mm) Pack of 5
10760631 Blue Tubes (16 mm) Pack of 5
10105830 Red Tubes (16 mm) Pack of 5
10503651 Yellow Tubes (16 mm) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
Product Code. 10603721 Supplier BRAND™ Supplier No. 4340000
Only null left
Add to basket
Add to basket
Constructed of sturdy polypropylene. Test Tube Racks include alphanumeric positions for easy sample identification. Racks are stackable, non-conductive, and will not float in water baths.
Sturdy polypropylene racks include alphanumeric positions for easy sample identification.
  • Racks are stackable, non-conductive and will not float in water baths.
  • Suitable for use in temperatures from -20° C to 90° C.
  • Supplied in three pieces for easy and permanent assembly.
  • Wide spaces between positions
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Color White
Holds Tubes (13 mm)
Material Polypropylene
Positions 84
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Dimensions (L x W x H) 265 x 126 x 75 mm
Length (Metric) 265 mm
Length (English) 10.43 in.
Width (English) 4.96 in.
Height (English) 2.95 in.
Diameter (Metric) Opening 13 mm
Product Type Test Tube Rack
Shelves None
Height (Metric) 75 mm
Array 6 x 14
Width (Metric) 126 mm