missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Pipet Tips, 2 to 200uL: Ultra-Low Retention
GREENER_CHOICE

Product Code. p-4901781 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
2 to 200 μL
Packaging:
Pk of 480
Pk of 960
Unit Size:
Pack of 480
Pack of 960
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Packaging unitSize
15194927 2 to 200 μL Pk of 480 Pack of 480
15184937 2 to 200 μL Pk of 960 Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15194927 Supplier BRAND™ Supplier No. 732308

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Low retention tips used for virtually all pipettes with yellow color code

  • High-purity polypropylene is oleamide- and diHEMDA-free
  • Nonsterile
  • All unopened packages are DNA-, RNase-, ATP- and endotoxin-free
  • Thin-walled for high translucency, easy ejection and low waste
  • Graduations at 20 and 100μL for quick volume check
  • Available in ecological rack-and-stack formats
  • Colorless and placed into yellow-colored tip tray

  • Produced through special, patented physicochemical process
  • Homogeneous, defect-free surface thus produced has low surface tension
  • Reduce sample loss and provide substantially higher reproducibility when working with critical media
  • For biological samples that contain detergents such as Triton™ X-100, SDS, Tween, etc.
  • High chemical resistance
  • Tips can be autoclaved at 121°C (2 bar) without damaging material properties

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 2 to 200 μL
Sterility Non-sterile
Color Colorless
Autoclavable Autoclavable
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) High translucency, easy ejection and low waste, quick volume check
Format Rack
Tip Style Low Retention
Packaging Pk of 480
Graduated Yes
Material Polypropylene
Graduations 20, 100μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.