Learn More
Fisherbrand™ Borosillicate Glass Funnel
Plain glass funnel manufactured from borosilicate glass. Fisherbrand™ Borosilicate Glass Funnel is ideal for use with funneling liquids in laboratory applications.
19.80€ - 47.85€
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Length (Metric) Stem | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Length (Metric) Stem | Price | Quantity & Availability | |||
11592423
|
Fisherbrand™
|
100mm | 12mm | 110mm |
41.00€
Pack of 10 |
In Stock
Log in to see stock. |
|||
11512433
|
Fisherbrand™
|
215mm | 24mm | 180mm |
47.85€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
11502433
|
Fisherbrand™
|
155mm | 19mm | 150mm |
19.80€
Each |
In Stock
Log in to see stock. |
|||
11572423
|
Fisherbrand™
|
55mm | 8mm | 60mm |
24.90€
Pack of 10 |
In Stock
Log in to see stock. |
|||
11582423
|
Fisherbrand™
|
75mm | 9mm | 80mm |
32.35€
Pack of 10 |
In Stock
Log in to see stock. |
|||
Description
Plain glass funnel manufactured from borosilicate glass.Specifications
Borosilicate Glass |