Learn More
Fisherbrand™ Borosilicate Glass Crystallizer without Spout
Use the Fisherbrand™ Borosilicate Glass Crystallizer without Spout to perform crystallization.
Brand:  Fisherbrand™
Additional Details : Weight : 0.09000kg
Product Code. 11893822
  • 16.30€ / Each
Estimated Shipment: 02-07-2024
Description
  • Without spout
  • Weight: 0.8kg
Specifications
Specifications
300 mL
Borosilicate Glass
Evaporating Dish