missing translation for 'onlineSavingsMsg'
Learn More

Bicine, 98.5 to 101.5% (Dry Basis), Ultrapure

Product Code. p-7082830 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15895088 100 g Plastic bottle 100g
15895258 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15895088 Supplier Thermo Scientific Supplier No. J12091.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 150-25-4 | C6H13NO4 | 163.173 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H13NO4
Molecular Weight (g/mol) 163.173
MDL Number MFCD00004295
InChI Key FSVCELGFZIQNCK-UHFFFAOYSA-N
Synonym N,N-bis[2-Hydroxyethyl]glycine
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES C(CO)N(CCO)CC(=O)O

Specifications

Melting Point ∼191°C (decomposition)
Physical Form Crystalline Powder
Quantity 100 g
Linear Formula (CH2CH2OH)2NCH2COOH
Packaging Plastic bottle
Beilstein 1769362
Formula Weight 163.17
Percent Purity 98.5 to 101.5%
Assay Dry basis
Chemical Name or Material Bicine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.