missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ BICINE, 0.5M buffer soln., pH 8.0

42.30€ - 66.00€

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H13NO4
Molecular Weight (g/mol) 163.173
MDL Number MFCD00004295
InChI Key FSVCELGFZIQNCK-UHFFFAOYSA-N
PubChem CID 8761
ChEBI CHEBI:40957
IUPAC Name 2-[bis(2-hydroxyethyl)amino]acetic acid
SMILES C(CO)N(CCO)CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15476079 SDS Alfa Aesar™
J63924.AE
100mL 42.30€
100mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-05-2019
Add to basket
 
15486079 SDS Alfa Aesar™
J63924.AK
250mL 66.00€
250mL View alternative products
Save Up to 
»
EU Stock 5
Add to basket
 
Description

Description

Applications
BICINE is used for the preparation of stable substrate solution for serum guanase determination. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

150-25-4
163.173
FSVCELGFZIQNCK-UHFFFAOYSA-N
CHEBI:40957
C(CO)N(CCO)CC(=O)O
C6H13NO4
MFCD00004295
8761
2-[bis(2-hydroxyethyl)amino]acetic acid

Specifications

BICINE buffer soln.
C6H13NO4
0.5M buffer soln.
C(CO)N(CCO)CC(=O)O
163.173
CHEBI:40957
8.0
150-25-4
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]acetic acid
8761
Liquid
100mL