Learn More
Invitrogen™ beta-2 Defensin Human ELISA Development Kit (TMB), PeproTech®, PeproTech®
ELISA
Brand:  Invitrogen™ 900-T172
Additional Details : Weight : 0.03500kg
Product Code. 17808965
  • 330.00€ / Pack of 10
Estimated Shipment: 11-06-2024
Description
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.
Specifications
Specifications
O15263
1673
10 x 96 Tests
Refer to product documentation for component specific storage temperature.
ELISA
DEFB4A