Learn More
Bel-Art™ Egg-Shaped Spinbar™ Magnetic Stir Bars
Product Code. p-166079
Change view
Click to view available options
Length (Metric):
9.5 mm
19 mm
25.4 mm
32 mm
38 mm
50.8 mm
76 mm
Diameter (Metric):
4.7 mm
9.5 mm
12.7 mm
16 mm
19 mm
19.1 mm
Length (English):
0.37 in.
0.75 in.
1 in.
1.25 in.
1.5 in.
2 in.
2.99 in.
Diameter (English):
0.18 in.
0.37 in.
0.5 in.
0.62 in.
0.75 in.
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Length (English) Diameter (English) unitSize
12358229 19 mm 9.5 mm 0.75 in. 0.37 in. Each
11316954 9.5 mm 4.7 mm 0.37 in. 0.18 in. Each
12398229 50.8 mm 19 mm 2 in. 0.75 in. Each
12368229 25.4 mm 12.7 mm 1 in. 0.5 in. Each
12378229 32 mm 16 mm 1.25 in. 0.62 in. Each
12388229 38 mm 16 mm 1.5 in. 0.62 in. Each
12308239 76 mm 19.1 mm 2.99 in. 0.75 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12358229 Supplier Bel-Art™ Supplier No. F371300034
Only null left
Add to basket
Add to basket
Designed for use in round-bottom vessels
  • Provide their own pivot point without a separate ring or abrupt change in contour for unrestricted spin
  • Minimal contact when used in plastic containers
  • Magnet itself is egg-shaped encapsulated in FDA grade, USP Class VI Teflon™ PTFE, except three smallest sizes have a cylindrical magnet in egg-shaped Teflon PTFE
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification
  • Autoclave safe
Specifications
Autoclavable Autoclavable
Certifications/Compliance FDA, USP Class VI, ISO 9001:2008, SGS
Color White
Diameter (English) 0.37 in.
For Use With (Equipment) 10, 25, 50 and 100mL round-bottom flasks
Length (English) 0.75 in.
Coating PTFE, FDA-grade, USP Class VI
Diameter (Metric) 9.5 mm
Length (Metric) 19 mm
Material Alnico Magnet
Quantity 1
Shape Egg
Type Magnetic Stir Bar