missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array Rat IL-10 Flex Set Standard
Lyophilized Rat IL-10
Brand: BD 558482
Additional Details : Weight : 0.01000kg
Description
- Lyophilized in an aqueous buffered solution containing BSA and ProClin™150
- Protein concentration of 25,000pg/ml (when reconstituted in 0.4mL Assay Diluent)
- Regulatory Status: RUO
- Recombinant
Flow cytometry, controls
Order Info
100 tests.
Specifications
| Aqueous Buffered Solution containing BSA and ProClin™ 150 | |
| Lyophilized | |
| 100 Tests | |
| RUO | |
| 4°C | |
| Rat IL-10 Flex Standard Set |
| Flow cytometry | |
| Technical data sheet | |
| 100000pg/mL | |
| Rat | |
| 0.2mL |