missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Mouse IL-4 Flex Set Standard

Standard control for flow cytometry

Brand:  BD 558477

Additional Details : Weight : 0.01000kg

Product Code. 15899418

  • 121.90€ / Each


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Description

Description

  • Contains lyophilized recombinant mouse interleukin-4
  • Size of 0.2mL

Specifications

Specifications

Aqueous Buffered Solution containing BSA and ProClin™ 150
Lyophilized
100 Tests
RUO
0.2mL
Flow cytometry
Technical data sheet
25000pg/mL
4°C
Videos

Research-use-only.