missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Mouse IFN-γ Flex Set Standard

Useful as a standard control with CBA Mouse IFN-γ Flex Set

Brand:  BD 558473

Additional Details : Weight : 0.01000kg

Product Code. 15859418

  • 121.90€ / Each


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Description

Description

  • Contains lyophilized recombinant mouse interferon-γ (IFN-γ)
  • When reconstituted in 0.4mL assay diluent, has a protein concentration of 25,000pg/mL

Specifications

Specifications

Aqueous Buffered Solution containing BSA and ProClin™ 150
Lyophilized
100 Tests
RUO
0.2mL
Flow cytometry
Technical data sheet
50000pg/mL
4°C
Mouse IFN-γ Flex Standard Set
Videos