missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Human MIP-1α Standard

Useful as a standard control with the CBA Human MIP-1a Flex Set

Brand:  BD 558468

Additional Details : Weight : 0.01000kg

Product Code. 15809418

  • 121.90€ / Each


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Description

Description

  • Contains lyophilized recombinant human macrophage inflammatory protein-1α
  • When reconstituted in 0.4mL assay diluent, has a protein concentration of 25,000pg/mL

Specifications

Specifications

Aqueous Buffered Solution containing BSA and ProClin™ 150
Lyophilized
100 Tests
RUO
0.2mL
Flow cytometry
Technical data sheet
25000pg/mL
4°C
Human MIP-1α Flex Standard Set
Videos