missing translation for 'onlineSavingsMsg'
Learn More

BD Cytometric Bead Array Human MIP-1β Standard

Lyophilized human MIP-1β

Brand:  BD 558469

Additional Details : Weight : 0.01000kg

Product Code. 15819418

  • 121.90€ / Each


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Description

Description

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Protein concentration of 25,000pg/ml (when reconstituted in 0.4ml Assay Diluent)
  • Regulatory status: RUO
  • Recombinant

Flow cytometry, controls

Order Info

100 tests.

Specifications

Specifications

Aqueous Buffered Solution containing BSA and ProClin™ 150
Lyophilized
100 Tests
RUO
4°C
Human MIP-1β Flex Standard Set
Flow cytometry
Technical data sheet
25000pg/mL
Human
0.2mL
Videos