missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array Human Interleukin-13 Flex Set Standard
Reconstituted Human IL-13 Standard is useful as a standard control with the CBA Human IL-13 Flex Set
Brand: BD 558460
Additional Details : Weight : 0.01000kg
Description
- Contains lyophilized recombinant human Interleukin-13
- When reconstituted in 0.4mL assay diluent, it has a protein concentration of 25,000pg/mL
Specifications
| Aqueous Buffered Solution containing BSA and ProClin™ 150 | |
| Lyophilized | |
| 100 Tests | |
| RUO | |
| Human IL-13 Flex Standard Set |
| Flow cytometry | |
| Technical data sheet | |
| 25000pg/mL | |
| 4°C |