Learn More
BD Cytometric Bead Array (CBA) Mouse IL-3; Flex Set, Bead A8
Immunoassay for measuring mouse interleukin-3 in serum and cell culture supernatant samples
Brand:  BD 558346
Additional Details : Weight : 0.10000kg
Product Code. 10378613
  • 865.00€ / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Includes: Mouse IL-3; Standard; Mouse IL-3; PE Detection Reagent; Mouse IL-3; Capture Bead A8
Description
Description
The BD CBA Mouse IL-3 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-3 (IL-3) in serum and cell culture supernatant samples.
  • Reactivity: Mouse
  • Reactivity was determined by testing samples with BD CBA Mouse IL-3 Flex set
Specifications
Specifications
IL-3
Mouse IL-3, standard, Mouse IL-3, PE detection reagent, Mouse IL-3, capture bead A8
Serum, Cell Culture Supernatant
Flow Cytometry
Mouse
100 Tests
4°C
Mouse IL-3 flex set (bead A8)