Learn More
BD Cytometric Bead Array (CBA) Mouse GM-CSF; Flex Set, Bead B9
A bead-based immunoassay for measuring mouse granulocyte-macrophage colony stimulating factor (GM-CSF) in serum and cell culture supernatant samples
Brand: BD
Additional Details : Weight : 0.10000kg
Includes: Mouse GM-CSF; Standard; Mouse GM-CSF; PE Detection Reagent; Mouse GM-CSF; Capture Bead B9
Description
Mouse reactivity is determined by testing samples with the BD CBA Mouse GM-CSF Flex Set.
- Reactivity: Mouse
- Storage Buffer: Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
Flow Cytometry
Specifications
GM-CSF | |
Mouse GM-CSF, standard, Mouse GM-CSF, PE detection reagent, Mouse GM-CSF, capture bead B9 | |
Serum, Plasma, Cell Culture Supernatant | |
Flow Cytometry |
Mouse | |
100 Tests | |
4°C | |
Mouse GM-CSF flex set (bead B9) |
Safety and Handling
- Mouse GM-CSF Flex Set
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : CMIT/MIT
Research Use Only