Learn More
BD Caspase-8 Inhibitor, Z-IETD-FMKK
Irreversible and cell permeable inhibitor of caspase-8
Brand: BD 550380
Additional Details : Weight : 0.01000kg
Description
Members of the caspase family play key roles in inflammation and mammalian apoptosis.The Z-IETD-FMK peptide is O-methylated in the P1 position on aspartic acid providing enhanced stability and increased cell permeability. The Z-IETD-FMK inhibitor is designed to be used in both in vivo and in vitro cell based assays to inhibit caspase-8 activity and to study events downstream of caspase-8 activation.
- Format: Purified
- Molecular weight: 654 Daltons
- Storage Buffer: Lyophilized in dimethyl sulfoxide (DMSO)
Avoid multiple freeze-thaws of product.
Fluorescence quantitation
Specifications
| Purified | |
| 654 | |
| Fluorescence quantitation | |
| -20°C |
| Z-IETD-FMK | |
| 1mg | |
| Lyophilized in Dimethyl Sulfoxide |
For research use only.