Learn More
BD Caspase-3 Inhibitor, Z-DEVD-FMK
Irreversible and cell permeable inhibitor of caspase-3
Brand: BD 550378
Additional Details : Weight : 0.01000kg
Description
Members of the caspase family play key roles in inflammation and mammalian apoptosis. The Z-DEVD-FMK peptide is O-methylated in the P1 position on aspartic acid providing enhanced stability and increased cell permeability. The Z-DEVD-FMK inhibitor is designed to be used in both in vivo and in vitro cell based assays to inhibit caspase-3 activity and to study events downsteam of caspase-3 activation.
- Contents: Lyophilized; Reconstitute in DMSO
- Format: Purified
- Molecular weight: 668 Dalton
- Storage buffer: Lyophilized in dimethyl sulfoxide (DMSO)
Avoid multiple freeze-thaws of product.
Flow cytometry
Specifications
| Purified | |
| 668 | |
| Flow cytometry | |
| -20°C |
| Z-DEVD-FMK | |
| 1mg | |
| Lyophilized in Dimethyl Sulfoxide |
For research use only.