missing translation for 'onlineSavingsMsg'
Learn More

BD Ac-DEVD-CHO Inhibitor

To identify and quantify Caspase-3 activity in apoptotic cells, and to study events downstream of Caspase-3 activation

Brand:  BD 556465

Additional Details : Weight : 0.01000kg

Product Code. 15828708

  • 132.90€ / Each


Estimated Shipment date
from Supplier 23-04-2019
Add to basket

Description

Description

Members of ICE/CED-3 cysteine protease family have key roles in inflammation and mammalian apoptosis. The ICE family member Caspase-3 (also known as CPP32, Yama, apopain) is activated early in apoptosis and appears to be involved in proteolysis of several important molecules, including poly (ADP ribose) polymerase (PARP)

  • Synthetic tetrapeptide inhibitor for Caspase-3
  • Used in protease assays
  • Cleaves PARP from 116kD to 85kD residual fragment
  • Cleavage site in PARP is C-terminal to Asp-216
  • Upstream sequence of cleavage site, DEVD (Asp-Glu-Val-Asp), is utilized as basis for highly specific Caspase-3 substrate, Ac-DEVD-AMC and Caspase-3 inhibitor,Ac-DEVD-CHO
  • Storage Buffer: Lyophilized powder
  • Format: Purified

Functional assay

Specifications

Specifications

Purified
502.5
Functional assay
-20°C
Ac-DEVD-CHO
1mg
Lyophilized powder
Videos