Learn More
Axygen™ Septa Mat for ABI multi-capillary Sequencing Instrument
Septa Mat for ABI Multi-Capillary Sequencing Instrument
Brand:  Axygen™ AM-96-SEPTA-3100
798.91 EUR valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Additional Details : Weight : 1.18000kg
Product Code. 12587897
  • 875.00€ / Pack of 50
In Stock
Description
Axygen™ AxyMatsTM septa mat is designed to be used on ABI sequencer. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
  • Septa Mat for ABI Multi-Capillary Sequencing Instrument
  • For use with 96 PCR microplates
  • Specifications
    Specifications
    ABI multi-capillary sequencing instruments
    Septa Mat